Learn More
LABboards Disposable Dissection Boards
Uses patented technology sterile and nonsterile boards for numerous applications including anatomic pathology dissection, surgical reconstructive surgery and tissue banking procedures
Manufacturer:  LAB BOARDS LLC 0711A
 View more versions of this product
Catalog No. 22-260-802
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Yes
Anatomic pathology dissection, surgical reconstructive surgery and tissue banking procedures
Poly Coated SBS Paper
Uses patented technology sterile and nonsterile boards for numerous applications
7 x 11 in.
Blank dissection board