Learn More
LABboards Disposable Dissection Boards
Uses patented technology sterile and nonsterile boards for numerous applications including anatomic pathology dissection, surgical reconstructive surgery and tissue banking procedures
Specifications
Material Poly Coated SBS Paper
For Use With (Application) Uses patented technology sterile and nonsterile boards for numerous applications
Includes Anatomic pathology dissection, surgical reconstructive surgery and tissue banking procedures
View More Specs
Products 1
Catalog Number Mfr. No. Type Size Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Type Size Quantity Price Quantity & Availability  
22-260-801
View Documents
LABboards
1218A
Metric dissection board 12 x 18 in. 50/Pk. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Prevents cross-contamination usually associated with multiple specimen types
  • Printed metric measurements provides accurate, hands-free specimen measurements
  • Durable, poly-coated SBS Paperboard or All-poly construction
  • Excellent operating environment for “wet skin” manipulation during procedure
  • Disposable cutting surfaces
Specifications
Specifications
Poly Coated SBS Paper
Uses patented technology sterile and nonsterile boards for numerous applications
Anatomic pathology dissection, surgical reconstructive surgery and tissue banking procedures