LABboards Disposable Dissection Boards Available on GSA/VA Contract for Federal Government customers only.
Uses patented technology sterile and nonsterile boards for numerous applications including anatomic pathology dissection, surgical reconstructive surgery and tissue banking procedures
Manufacturer: LAB BOARDS LLC 0912A
 View more versions of this product
Catalog No. 22-260-800
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Disposable Yes
    For Use With (Application) Uses patented technology sterile and nonsterile boards for numerous applications
    Includes Anatomic pathology dissection, surgical reconstructive surgery and tissue banking procedures
    Size 9 x 12 in.
    Material Poly Coated SBS Paper
    Type Metric dissection board
    Product Type Metric dissection board
  • Description & Specifications
Related Products
Product Content Feedback