Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DenLine™ Protection Plus+™ II Unisex Long-Length Lab Coats, Color: White

Provides 360 deg. employee protection and comfort needs with high breathable front and back fabric.

Manufacturer:  DenLine™ DL270BJWHT4XL

 View more versions of this product

Catalog No. 22-412-351


Add to cart

Description

Description

  • Fluid Resistance: Incidental Splash
  • Stain resistant to all aqueous liquids like blood, bleach, select reagents
  • Lint free fabric; Clean Room Level ISO 5 (Class 100)
  • Three pockets
  • Double stitched
  • Bar Tack reinforced
  • Side vents
  • Tapered sleeves prevents catching on things when reaching
  • Spun woven, low pill, “Quick Dry” cuffs
  • Heavy duty stainless steel snaps (lifetime guaranteed)
  • For bloodborne applications

Specifications

  • Length: 104cm (41 in.)
  • Hydrostatic Pressure (AATCC 127): >200mm
  • Air Permeability (ATSM D 737): 40 cu. ft./min.
  • Breathability: 50% Poly/50% Cotton
  • Static Resistance: Surface Resistivity 109 (Test Method 76)

    Wash Life

    • 200+ Wash Life - Commercial Wash Tested through 225 Hot Water Wash Cycles, Meets CDC HICPAC Recommendations for Processing Medical Linen.
Specifications

Specifications

Snaps
99% polyester and 1% carbon
No
Unisex
104 cm
3
Incidental splash, stain resistant to all aqueous liquids like blood, bleach, select reagents
58 to 60 in.
28 to 30 in.
137 to 146 cm
Long-Length
White
Knit
For bloodborne applications
41 in.
Polycotton blend
Long-Length Lab Coat
4X-Large
54 to 57.5 in.
147 to 152 cm
Tapered
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit