Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinets, 6 ft. W, 8 in. Opening

Features ergonomic Inclination™ Technology and MyLogic™ Operating System

Manufacturer:  Labconco™ 304681120

 View more versions of this product

Catalog No. 30-468-1120


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

32.6 in.
100V, 50/60Hz, 12A, International
245.1cm
Fluorescent
Safety Cabinet
1000 lb.
78.2 in.
NSF International-Listed, ETL, Modified ASHRAE 110
20.3cm
82.8cm
Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
UV light, service fixture, Vacu-Pass Portal, Base stand
Japan
8 in.
453.6kg
198.6cm
96.5 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit