Labconco™ Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinets, 6 ft. W, 8 in. Opening

Features ergonomic Inclination™ Technology and MyLogic™ Operating System

Manufacturer: Labconco™ 304680060

 View more versions of this product

Catalog No. 30-468-0060

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart

    Specifications

    Depth (English) 32.6 in.
    Depth (Metric) 82.8cm
    For Use With (Application) Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
    Product Type Safety Cabinet
    Height (English) 69 in.
    Electrical Requirements 230V, 50/60Hz, 6A, International
    Height (Metric) 175.2cm
    Plug Type British (UK, EN Compliant)
    Width (English) 78.2 in.
    Width (Metric) 198.6cm
    Certifications/Compliance NSF International-Listed, ETL, Modified ASHRAE 110
    Weight (Metric) 408.2kg
    Sash Opening (Metric) 20.3cm,20.3cm
    Lighting Fluorescent
    Weight (English) 900 lb.
    Includes N/A
    Sash Opening (English) 8 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit