Labconco™ Acid-Resistant Metal Rotors for CentriVap™ Vacuum Concentrators

Easily interchangeable PTFE-coated aluminum rotors with PTFE hubs hold tubes at an angle to maximize surface areas

Manufacturer: Labconco™ 7455201

 View more versions of this product

Catalog No. 16-108-451

  Check Availability
Add to Cart

    Specifications

    Holds 32 x 1.5 to 2mL microcentrifuge tube or up to 12 x 40 to 50mL tube
    Capacity (Metric) 1.5 to 2mL or 40 to 50mL
    For Use With (Equipment) CentriVap™ vacuum concentrator
    Product Type Acid Resistant Rotor
    Material PTFE Coated Aluminum
    Weight (English) 5 lb.
    Weight (Metric) 2.3kg
    Total Count 32.0

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit