Labconco™ Acid-Resistant Metal Rotors for CentriVap™ Vacuum Concentrators

Easily interchangeable PTFE-coated aluminum rotors with PTFE hubs hold tubes at an angle to maximize surface areas

Manufacturer: Labconco™ 7450701

 View more versions of this product

Catalog No. 16-108-448

  Check Availability
Add to Cart

    Specifications

    Holds 132 x 1.5 to 2mL microcentrifuge tube
    Weight (English) 5 lb.
    Weight (Metric) 2.3kg
    Capacity (Metric) 1.5 to 2mL
    For Use With (Equipment) CentriVap™ vacuum concentrator
    Product Type Hexagonal Rotor
    Material PTFE Coated Aluminum
    Total Count 132.0

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit