Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Basic™ Laboratory Hood

For areas with low ceilings or narrow aisles, choose the Labconco Basic™ Laboratory Hood when outfitting your lab.

Manufacturer:  Labconco™ 2247500

 View more versions of this product

Catalog No. 16-080-1


  Check Availability
Add to cart

Requires : Remote blower (some models), ductwork, base cabinets, adjustable-height base stands, solid epoxy flat work surfaces.

Specifications Description & Specifications

Specifications

25 in.
53 in.
47 in.
Integral explosion-proof blower included
166kg
Glacier White
115/208-230V, 60Hz, 7.8/4.4A, Domestic/International
63.5cm
134.6cm
119.4cm
365 lb.
ANSI Z9.5, ASHRAE 110, ASTM E84, CAN/CSA C22.2, CE, NFPA 45, SEFA 1, UL
Explosion-proof incandescent
Laboratory Hood
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit