Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Benchtop FreeZone™ Legacy™ Freeze-Dry Systems, 208/230V 60Hz Models: 6L

With small footprint and four feet, this smaller capacity model fits easily on benchtop and is ideal for light and moderate loads

Manufacturer:  Labconco™ 7758040

 View more versions of this product

Catalog No. 10-274-57


  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Add to cart

Requires : Required Accessories
  • Drying chambers or other drying accessories, vacuum pumps and glassware
  • Require drying accessories


Specifications Description & Specifications

Specifications

6L
208/230V 60Hz
60Hz
80.8cm
≤4L
71.4cm
31.875 in.
208/230V
31.875 x 28.0625 x 14.625 in. (80.8 x 71.4 x 37.1cm)
37.1cm
Stainless-steel coil and collector chamber, Stoppering Tray Dryer
Benchtop Freeze-Dry System
28.0625 in.
14.625 in.
-50°C
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit