Learn More
Labconco™ Carbon Exhaust Flow Filters for Trace Organics
Manufacturer:  Labconco™ 3937201
 View more versions of this product
Catalog No. 10-304-140
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Filter
45.7cm
18 in.
2 ft. Purifier™ class I enclosures, Purifier™ HEPA filtered enclosures, XPert™ filtered balance system
Formaldehyde
2.5cm
3.4kg
10 to 20 x 18 x 1 in. (25.4 to 50.8 x 45.7 x 2.5cm)
18 in.
45.7cm
Carbon
10 to 20 in.
7.5 lb.
1 in.
25.4 to 50.8cm