Learn More
Labconco™ Ceiling Enclosure Kits for Protector™Premier™, XL and XStream™ Laboratory Hoods
Ceiling enclosures extend top of hood to the ceiling to hide exposed ductwork, plumbing and wiring
Supplier: Labconco™ 9414401
Description
- Fit any 3 - 8 ft. Protector Premier, XL or XStream Laboratory Hood (based on appropriate hood depth and nominal width)
- Each kit includes three ceiling enclosure panels — one for front of hood and one for each side of the hood
- Panels are constructed of white, powder-coated steel
Specifications
Protector™ Premier™, XStream™, XL™ fume hood | |
48 in. | |
121.9 cm | |
80.52 cm |
14 to 18.6 in. | |
35.56 to 47.24 cm | |
Protector™ Premier™, XStream™, XL™ fume hood | |
31.7 in. |