Learn More
Labconco™ CentriVap™ Complete Concentrator
Comes with everything needed to start processing
Manufacturer: Labconco™ 7315031
Description
Comes with everything needed to start processing
Uses a combination of centrifugal force, vacuum and heat to speed evaporation.
Includes:
- Vacuum Concentrator
- -50°C Cold Trap
- Aluminum rotor holds 40 microcentrifuge tubes and 12-13mm tubes
- Chemical-resistant diaphragm vacuum pump
- CentriZap Strobe Light allows user to view samples while rotor is spinning and lid is closed
Molecular biology, proteomics, genomics, cell biology, microbiology, biochemistry, drug discovery and analytical chemistry
Specifications
International | |
230V 50Hz, 6A | |
Centrivap | |
Acid-resistant, no heat boost |