Learn More
Labconco™ Class II Cabinets IV Bar Kit
SDP
Bar supports intravenous solution bottles and bags in Purifier Biosafety Cabinets.
Specifications
Product Type Exhaust Safety Cabinets Kit
View More Specs
Products 4
Catalog Number Mfr. No. Weight (English) Weight (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Weight (English) Weight (Metric) Price Quantity & Availability  
10-269-251
SDP
View Documents
Labconco™
3858601
1.81 kg
Each for $264.00
N/A
10-269-253
SDP
View Documents
Labconco™
3858603
5 lb. 2.26 kg
Each for $373.00
N/A
10-269-250
SDP
View Documents
Labconco™
3858600
3 lb. 1.36 kg
Each for $204.00
N/A
10-269-252
SDP
View Documents
Labconco™
3858602
6 lb. 2.72 kg
Each for $282.00
N/A
Description
Description
Bar supports intravenous solution bottles and bags. Kit includes IV bar, mounting hardware and four hangers. For use with Labconco Purifier Class II Biosafety Cabinets. Installation is required.
Specifications
Specifications
Exhaust Safety Cabinets Kit