Labconco™ Clear Stoppering Chambers

For use with 6L, 12L and 18L Freezone™ Dryers

Manufacturer: Labconco™ 7868020

 View more versions of this product

Catalog No. 10-030-164

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart

    Specifications

    Product Type Clear Stoppering Chamber
    Diameter (English) 13.5 in.
    Diameter (Metric) 34cm
    Electrical Requirements 115V 50/60Hz
    For Use With (Application) To provide economical means of stoppering serum bottles
    For Use With (Equipment) For 6l, 12l and 18l freezone™ dryers
    Height (English) 23 in.
    Height (Metric) 58.4cm
    Hertz 50/60Hz
    Weight (English) 30 lb.
    Weight (Metric) 13.6kg
    Voltage 115V

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit