Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Dished Work Surface

Labconco Dished Work Surface are contoured and designed to support a variety of Labconco enclosures.

Manufacturer:  Labconco™ 3909908

 View more versions of this product

Catalog No. 10-030-234


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Without Cutout
90cm
1 in.
Spills
155 lb.
60 in.
Black
Dished Work Surface
35.5 in.
Paramount™ and Paramount 360™ ductless enclosure, Purifier™ Class I enclosure, Purifier™ filtered PCR enclosure, Purifier™ HEPA filtered enclosure, Purifier
2.5cm
Chemical resistant
70kg
152cm
Solid Epoxy
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit