Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco Duplexes for Basic Hoods
For Labconco Basic 47 and 70 Fume Hoods
$47.25 - $110.00
Specifications
Amperage | 20A |
---|---|
For Use With (Application) | Mounting lower right-side panel of Basic™ hood |
For Use With (Equipment) | Basic™ hood |
Voltage | 115V |
Description
Duplex Electrical Receptacle Kits for use with Labconco Basic 47 and Basic 70 Fume Hoods. Each kit includes duplex electrical receptacle, junction box and receptacle cover plate. For mounting on the lower right side fixture panel. Electrical wiring is not included.Specifications
20A | |
Mounting lower right-side panel of Basic™ hood | |
Basic™ hood | |
115V |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title