Learn More
Labconco™ Exhaust Transition Adapters for XPert™and Protector™XVS
SDP
Exhaust duct transition adapters for ductwork on Labconco Enclosures.
Manufacturer:  Labconco™ 3912400
 View more versions of this product
Catalog No. 10-304-75
Add to cart
Description
Description
Exhaust duct transition adapters for ductwork on Labconco Enclosures.
Upper Exhaust Transition Adapter made of one-piece molded polyethylene. Connects 2 in. x 10 in. rectangular to 5 in. diameter round for connection to 5 in. ID hose (included with FilterMate Portable Exhauster). For use with Labconco XPert Enclosures, Protector XVS Ventilation Stations, Protector Demonstration Hoods, and Protector Work Stations.
Specifications
Specifications
Upper connection, 5 in. Hose
2.3kg
5 lb.
24, 36, 48 in. XPert™ and Protector™ XVS work station
10 in.
5 in.
Epoxy Coated Steel
Provide connection between the XPert™ enclosures and ductwork