Learn More
Labconco Exhaust Transition Adapters for XPertand ProtectorXVS
Exhaust duct transition adapters for ductwork on Labconco Enclosures.
Manufacturer: Labconco 3912402
Description
Exhaust duct transition adapters for ductwork on Labconco Enclosures.
Lower Exhaust Transition Adapter made of one-piece molded polyethylene. Connects 2 in. x 10 in. rectangular to 5 in. diameter round for connection to 5 in. ID hose. For use with Labconco XPert Enclosures, Protector XVS Ventilation Stations, Protector Demonstration Hoods, and Protector Work Stations.Specifications
Provide connection between the XPert™ enclosures and ductwork | |
5 lb. | |
5 in. | |
Epoxy Coated Steel |
24, 36, 48 in. XPert™ and Protector™ XVS work station | |
2.3kg | |
10 in. |