Learn More
Labconco™ Fast-Freeze™ Flasks, Complete Assembly
Easy to handle and fast to load
Manufacturer:  Labconco™ 7541200
 View more versions of this product
Catalog No. 10-269-56
  Check Availability
Add to cart
Requires : Adapters are required to attach flasks to freeze dry valves–40 and 80mL flasks use a 0.5 in. (1.3cm) adapter; larger sizes use a 0.75 in. (1.9cm).
Specifications Description & Specifications
Specifications
3.55 in.
90.2mm
14.9 in.
2.2 lb.
2000mL
1.9cm
Compatible with all major brands of freeze-drying equipment
38cm
1kg
Borosilicate Glass
Accessories