Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Fiberglass Motor Blowers

Ideal for fume hood exhaust systems in moderate to highly corrosive conditions

Manufacturer:  Labconco™ 7181000

 View more versions of this product

Catalog No. 16-090-35


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

0.4hp
4.4A
>1000 cfm
60Hz
569-753rpm
115V
44kg
Fiberglass
115V 60Hz
Fume hood exhaust system
569-753rpm
Blower
96 lb.
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit