Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ FilterMate™ Exhauster Filter Cells
Replacement filters for use with Labconco FilterMate Portable Exhausters.
Manufacturer: Labconco™ 3923402
Description
Ammonia Carbon Filter, Type CX, contains a minimum of 16 pounds of impregnated carbon media suitable for ammonia and low molecular weight amines.Specifications
18 in. | |
10 to 20 x 18 x 2.1 in. (25.4 to 50.8 x 45.7 x 5.3cm) | |
Ammonia and low molecular weight amines | |
2.1 in. | |
10 to 20 in. | |
Filter | |
9.5kg | |
45.7cm |
45.7cm | |
Carbon | |
FilterMate™ portable exhauster | |
5.3cm | |
25.4 to 50.8cm | |
21 lb. | |
18 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title