Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ FilterMate™ Portable Exhauster

Labconco FilterMate™ Portable Exhauster removes hazardous powders, particulates or vapors from exhaust air stream, returning filtered air to environment.

Manufacturer:  Labconco™ 3970023

 View more versions of this product

Catalog No. 10-304-38


  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Add to cart

Requires : Carbon Filter (Models for use with carbon filter only). Models for use with carbon filter require Organic Vapor Cell, Formaldehyde Impregnated Cell, or Ammonia Impregnated Cell. Combination models can support use of two carbon filters by removing HEPA filter.

Specifications Description & Specifications

Specifications

14.9 x 23 x 22.7 in. (38 x 58 x 58cm)
New_Flow Rate
12.7cm
1.2 ft.
250 cfm
14.9 in.
Ammonia-Amine, Formaldehyde, HEPA, Organic Vapor
38cm
50 lb.
58cm
CE
230V 50Hz
5 in.
Exhauster
38cm
22.7 in.
23 in.
50Hz
230V
22.7kg
58cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit