Learn More
Labconco™ FilterMate™ Portable Exhausters
Remove a broad range of contaminants and particulates
Manufacturer:  Labconco™ 3970001
 View more versions of this product
Catalog No. 10-304-33
  Check Availability
Add to cart
Requires : Cat. No. 10-304-35 requires outside ducting.
Specifications Description & Specifications
Specifications
250 cfm
23 in.
None, room exhaust
Carbon
Portable Exhauster
50 lb.
14 in.
CAN/CSA C22.2, UL
58cm
99.99%
Demonstration hood, Protector™ work station, Protector™ XVS ventilation station, XPert™ balance enclosure
115V
22.7kg
36cm