Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ FlaskScrubber™ Glassware Washer

Provide thorough, efficient, and economical cleaning that meets demanding laboratory standards by outfitting your lab with a Labconco FlaskScrubber™ Glassware Washer.

Manufacturer:  Labconco™ 4420421

 View more versions of this product

Catalog No. 10-359-113


  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Add to cart

Specifications Description & Specifications

Specifications

12A
230V 50/60Hz
34.2 to 36.2 in.
One spindle rack
120°F
93°C
60°C
20 to 120psi for hot tap water pressure at washer
Freestanding FlaskScrubber™
239 lb.
24.2 in.
UL 61010-1, CAN CSA C22.2 No. 61010.1, ETL, CE
69.9cm
50/60Hz
Domestic/international
4.7L/min. for hot tap water, 3.4L/min. for purified water
86.9 to 91.9cm
199°F
140°F
49°C
Type 304 Stainless Steel
0psi
13L/Fill
108kg
61.5cm
27.5 in.
Ideal for college and university, government, pharmaceutical and industrial labs or any facility utilizing reusable glassware
230V
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit