Learn More
Labconco™ FlaskScrubber™ Glassware Washer
Provide thorough, efficient, and economical cleaning that meets demanding laboratory standards by outfitting your lab with a Labconco FlaskScrubber™ Glassware Washer.
Manufacturer:  Labconco™ 4578130
 View more versions of this product
Catalog No. 10-359-187
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
16A
115V 60Hz
32.9 to 34.9 in.
One spindle rack, glass viewing window, 25w interior light
120°F
82°C
60°C
20 to 120psi for hot tap water pressure at washer
Undercounter Washer with viewing window and lighting
206 lb.
24.1 in.
UL 61010-1, CAN CSA C22.2 No. 61010.1, ETL
69.6cm
60Hz
Domestic
4.7L/min. for hot tap water, 3.4L/min. for purified water
83.6 to 88.6cm
180°F
140°F
49°C
Type 304 Stainless Steel
0psi
13L/Fill
93kg
61.2cm
27.4 in.
Ideal for college and university, government, pharmaceutical and industrial labs or any facility utilizing reusable glassware
115V
Promotions
Promotions
  • Labconco CorporationSupplier Diversity Partner
    Purchase any Labconco™ FlaskScrubber™ or SteamScrubber™ Glassware Washer and receive both a Utensil Basket and Utensil Holder at no additional cost. This is an NLSU exclusive offer. You must apply and qualify for the program to be eligible.
    06-30-2018