Learn More
Labconco™ Flexi-Bin Cart
The Labconco Flexi-Bin Cart provides for safe transport of glassware and chemical containers.
Manufacturer:  Labconco™ 8010000
 View more versions of this product
Catalog No. 16-317-74
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Flexi-bin cart
36.38 in.
34.88 in.
19 in.
34.88 x 19 x 36.38 in. (88.6 x 48.3 x 92.4cm)
181kg
Safe transport of Glassware and chemical containers
44.09 lb.
Steel
92.4cm
88.6cm
48.3cm
400 lb.
2
18G Welded 1 in. white tubular Steel frame, Rubber handle grips, removable 4 in. D Polyethylene bin, three long (20.25 in.) and four short (15.25 in.) Polyethylene dividers, 4 in. dia. casters, two with toe locks, ID tag
20kg