Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ FreeZone™ Console Freeze Dry System, 12 Liters

Designed for moderate aqueous sample loads

Manufacturer:  Labconco™ 7754044

 View more versions of this product

Catalog No. 16-208-5014


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

9A
12L
28 in.
230V 60Hz
36.3 in.
60Hz
Stainless Steel, Epoxy-coated Steel Exterior
Freeze Dry System
278 lb.
3.17 gal., 12L
3.17 gal.
CAN/CSA C22.2, UL
71.1cm
For moderate aqueous sample loads
92.2cm
Ptfe coated collector, purge valve
North America
Console
126kg
230V
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit