Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ FreeZone™ Console Freeze Dry System, 12 Liters

Designed for moderate aqueous sample loads

Manufacturer:  Labconco™ 7759041

 View more versions of this product

Catalog No. 16-208-5017

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Specifications

8, 9A
12L
31 in.
230V 60Hz
63.4 in.
60Hz
Stainless Steel, Epoxy-coated Steel Exterior
Freeze Dry System
697 lb.
3.17 gal., 12L
3.17 gal.
CAN/CSA C22.2, UL
78.7cm
For moderate aqueous sample loads
161cm
Ptfe coated collector, stoppering tray dryer
North America
Console
316kg
230V
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit