Labconco™ Fume Hood Standard Service Fixture Kits (Fiberglass 30 or Basic Hoods)

Keep your Labconco fume hood in top operation with a Fume Hood Standard Service Fixture Kit, available in several types to meet your specific needs.

Manufacturer: Labconco™ 2832605

 View more versions of this product

Catalog No. 16-331

  Check Availability
Add to Cart

    Specifications

    Product Type Gas Standard Service Fixture Kit
    For Use With (Equipment) Basic laboratory hoods, Fiberglass 30 laboratory hoods
    Pressure 40psi
    Weight (English) 4 lb.
    Flow Rate 29 cfm
    Includes Remotely controlled service fixture with 0.25 in. dia. brass valve and tubing, blue fixture knob and blue hose connector, for mounting on right side of fume hoods
    Weight (Metric) 1.8kg

    • Remote controlled
    • Right-side mount
    • Fits Fiberglass 30 or Basic hoods
    • Available types: Air, Argon, Cold Water, Deionized/Distilled Water, Gas, Hot Water, Nitrogen, Steam, Vacuum

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit