Learn More
Labconco™ G4 Prefilter Starter Kits
For use with Purifier™ Logic™-plus biosafety cabinets
Supplier: Labconco™ 3406001
Description
- Includes pleated disposable filter with Merv 8 rating, hardware and bracket
Installation is required.
Specifications
Prefilters and Postfilters | |
Pleated disposable filter with Merv 8 rating, hardware and bracket | |
4 lb. |
Purifier Logic and Biosafety Cabinet, 4 ft. | |
G4 Prefilter Starter Kit | |
1.8 kg |