Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Gas and Vacuum 0.2μm Filter

Traps particulates from drying train or vacuum pump exhaust

Manufacturer:  Labconco™ 5240100

Catalog No. 16-108-422


Add to cart

Description

Description

  • 0.25 to 0.38 in.
  • For use with all glove boxes

Specifications

Specifications

Enclosures, Precise™ Basic™ glove box, Precise™ controlled atmosphere glove box, Precise™ HEPA filtered glove box, Protector™ combination glove box
Filter
5 lb.
2.3kg
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit