Learn More
Labconco™ Glove Box Filters
For Labconco Filtered Glove Boxes
$1013.00 - $1907.00
Description
16-101-11
- Carbon Exhaust
- Use as optional second outlet filter for removal of gaseous organic contaminants in low level radioisotope or chemical carcinogen applications
- HEPA
- Inlet or exhaust
- Use as replacement filter or optional second outlet filter for removal of 0.3micrometer particulate contaminants, For Labconco Multi-Hazard Glove Boxes
Specifications
Multi Hazard Glove Box |