Learn More
Labconco™ Glove Box Filters
For Labconco Filtered Glove Boxes
Supplier: Labconco™ 5065300
Description
16-101-11
- Carbon Exhaust
- Use as optional second outlet filter for removal of gaseous organic contaminants in low level radioisotope or chemical carcinogen applications
- HEPA
- Inlet or exhaust
- Use as replacement filter or optional second outlet filter for removal of 0.3micrometer particulate contaminants, For Labconco Multi-Hazard Glove Boxes
Specifications
Multi Hazard Glove Box | |
10 lb. | |
4.5 Kg |