Labconco™ Guardian™ Digital Airflow Monitor: For Premier, Protector
Monitor and alert operator to low airflow conditions
Manufacturer: Labconco™ 9413400
 View more versions of this product
Catalog No. 10-369-206
  Check Availability
Add to Cart
    Specifications
    Hertz 50/60Hz
    For Use With (Equipment) Premier, Protector XL, Protector Xstream lab hoods
    Voltage 100-115V
    Electrical Requirements 100-115V 50/60Hz
  • Description & Specifications
Related Products
Product Content Feedback