Learn More
Labconco™ Guardian™ Jr. Airflow Monitor Kits
Monitor and alert the operator to low airflow conditions
Supplier: Labconco™ 9743204
Includes: Airflow sensor, faceplate with circuit board, powder-coated steel housing, electrical cord with power pack and plug, vinyl tubing, airflow tube adapter and mounting hardware
Description
- From faceplate, easily select and calibrate a setpoint between 50 and 230fpm using a velocity meter and a screwdriver
- If hood face velocity falls below programmable setpoint for more than six seconds, audible alarm sounds and ALERT indicator glows red
- TEST RESET switch mutes audible alarm
Order Info
Installation requires factory-prepared cut-out in upper portion of front panel (for ClassMate) or right fixture panel (for other models).
Specifications
Protector™ ClassMate™ Hood | |
2.7 Kg | |
115 V | |
60 Hz |
6 lb. | |
Fume Hood Accessory | |
115 V, 60 Hz |