Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Labconco™ HEPA Filter for Rotary Vane Vacuum Pump and FreeZone™ Freeze Dry Systems
Fits between freeze dryer and pump to prevent entry of particulate contaminants into pump
Manufacturer:  Labconco™ 7386500
Catalog No. 09-040-130
Description
Description
HEPA Filter for all Labconco FreeZone Freeze Dry System when connecting to Labconco Rotary Vane or Hybrid Vacuum Pumps. Filter fits between the freeze dryer and pump to prevent particulate contaminants from getting into the pump.
Specifications
Specifications
1 lb.