Learn More
Labconco™ HEPA Filter
SDP
For Purifier Class I and HEPA-Filtered Enclosures and XPert and RXPert Enclosures.
Supplier:  Labconco™ 3707902
Catalog No. 10-304-146
Add to Cart
Description
Description
Supply HEPA filter for 4' Purifier Class I Enclosures, Purifier Filtered PCR Enclosures, Purifier HEPA Filtered Enclosures, Purifier Vertical Clean Benches, XPert Filtered Balance Stations, XPert & RXPert Filtered Balance Systems.
Specifications
Specifications
99.99%
4 ft. Purifier™ class I safety, HEPA filtered, vertical clean bench and PCR enclosure
71.7 Kg
101.6 to 127 cm
42 in.
3.3 in.
HEPA
158 lb.
40 to 50 in.
18 in.
Particulate