Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Manual and Electric Hydraulic Lift Base Stands

Offers infinitely adjustable height from 25.5 and 33.5 in. (648 to 851mm)

Manufacturer:  Labconco™ 3780102

 View more versions of this product

Catalog No. 09-040-133


Add to cart

Description

Description

  • Height adjustment on manual unit can be done via hand crank. Height adjustment on electric units using up/down switch
  • Fixed feet
  • Available width rage: 3 to 6 ft.

When installing the Cabinet on the hydraulic lift base stand, ensure that the hydraulic lines and the electrical cord are clear of any obstructions before installing the cabinet on the stand or operating the lift system.

Specifications

Specifications

34.3 in.
189.2cm
Electric Hydraulic Lift
ADA, NSF 49, UL
210 lb.
87.1cm
75.7 in.
115V
34.5 to 25.5 in.
95kg
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit