Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Motor Blowers: Coated Steel

For low to moderately corrosive applications

Manufacturer:  Labconco™ 7068300

 View more versions of this product

Catalog No. 16-090-14


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

0.25hp
4.5A
Moderately corrosive applications
60Hz
115V
42kg
25.4cm
Coated Steel
115V 60Hz
Protector™ Premier™, Protector™ XStream™, Basic™ 47 and 70 and fiberglass 30 fume hood
Blower
92 lb.
10 in.
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit