Learn More
Labconco™ Paramount™ Ductless Enclosure Filters
Filters for Paramount Ductless Enclosures
$363.00 - $590.00
Specifications
For Use With (Equipment) 6 ft. Paramount™ ductless enclosure
Product Type Filter
View More Specs
Products 5
Catalog Number Mfr. No. Filter Type For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Filter Type For Use With (Application) Price Quantity & Availability  
10-030-224
View Documents
Labconco™
6938200
Carbon Organic Vapor 7.5#
Each for $363.00
Add to cart
10-030-225
View Documents
Labconco™
6938201
Carbon Acid-Sulfur 10#
Each for $562.00
Add to cart
10-030-228
View Documents
Labconco™
6938204
Carbon Mixed Bed (Org-Acid-Ammon-Form) 10#
Each for $514.00
Add to cart
10-030-227
View Documents
Labconco™
6938203
Carbon Formaldehyde-Formalin 10#
Each for $590.00
Add to cart
10-030-230
View Documents
Labconco™
6938100
HEPA Particulate
Each for $506.00
Add to cart
Description
Description
For use with Paramount Ductless Enclosures
Order Info
Acid gas detector tubes available. (See Cat. No. 10-030-241)
Specifications
Specifications
6 ft. Paramount™ ductless enclosure
Filter