Labconco™ Pressure Gauge Kits for Precise™ and HEPA-Filtered Glove Boxes
Accessories for use with Precise Glove Boxes and HEPA-Filtered Glove Boxes, XPert Weigh Boxes
Manufacturer: Labconco™ 5238100
 View more versions of this product
Catalog No. 16-108-420
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Type Transfer chamber pressure gauge kit
    For Use With (Equipment) Precise™ HEPA filtered glove box, XPert™ weigh box
    For Use With (Application) Monitors pressures between 0 to 29 in. of mercury
    Weight (English) 2 lb.
    Weight (Metric) 0.9kg
  • Description & Specifications
Related Products
Product Content Feedback