Learn More
Labconco™ PrimeMate™ Drain/Fill Adapter
For use with Labconco PrimeMate Oil Changing System
Manufacturer: Labconco™ 7358700
Requires : PrimeMate Quick Disconnect Coupler (Cat. No. 10-269-742)
Description
- Used to attach drain/fill coupler to vacuum pumps with 7/8-14 drain plug threads
- Weight: 0.2kg (.5 lbs.)
Specifications
PrimeMate Oil Change System | |
Drain/Fill Tubing Adapter |