Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Protector™Acid Storage Cabinets

Support Labconco fume hoods and safely store and vent acids and other corrosive liquids

Manufacturer:  Labconco™ 9905000

 View more versions of this product

Catalog No. 16-080-306


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

ADA
55.88cm
Ideal for storage and venting of small laboratory instrument and mild acid and other reagent
80 to 83.18cm
800 lb.
Steel
120 lb.
54.43kg
22 in.
Single Right Hinged Door
31.5 to 32.75 in.
Vacuum formed PVC liner tray, 2 rear vent connection cut outs for venting the cabinet into a fume hood, 4 leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30in.
362.8kg
Acid Storage Cabinet
24 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit