Labconco™ Protector™Acid Storage Cabinets

Support Labconco fume hoods and safely store and vent acids and other corrosive liquids

Manufacturer: Labconco™ 9905100

 View more versions of this product

Catalog No. 16-080-307

  Check Availability
Add to Cart

    Specifications

    Certifications ADA
    Height (English) Base 31.5 to 32.75 in.
    Height (Metric) Base 80 to 83.18cm
    Load Capacity (English) Centralized 800 lb.
    Material Steel
    Width (English) 18 in.
    Depth (Metric) 55.88cm
    Door Type Single Right Hinged Door
    For Use With (Application) Ideal for storage and venting of small laboratory instrument and mild acid and other reagent
    Includes Vacuum formed PVC liner tray, 2 rear vent connection cut outs for venting the cabinet into a fume hood, 4 leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30in.
    Load Capacity (Metric) Centralized 362.8kg
    Product Type Acid Storage Cabinet
    Weight (English) 85 lb.
    Weight (Metric) 38.55kg
    Depth (English) 22 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit