Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ Fume Hood Accessory, SpillStopper™ Work Surface
SpillStopper Work Surface is designed for protecting your Labconco Protector™ Echo Filtered, Airo Filtered or XL Fume Hood from accidental spills.
Manufacturer: Labconco™ 9501622
Description
- Molded from special formulation of corrosion-resistant epoxy resin
- Dished surface to contain spills
- Contoured to conform to interior liner of Protector Echo, Airo and XL hoods
- Pre-drilled 5.8cm (1.5 in.) diameter holes for venting
- Pre-drilled 6.4cm (2.5 in.) for plumbing pass-through
- Front edge has large radius to aerodynamically direct airflow into hood
- May be ordered with a pre-cut 15.2 x 7.6cm (6 x 3 in.) oval cupsink cutout(s) in left rear, right rear, left side, or right side
- Cupsink sold separately
Cupsink cutouts are not for use with Protector Solvent Storage Cabinets
Specifications
36 in. | |
91.4cm | |
6 ft. Protector Echo Filtered, Airo Filtered or XL Fume Hood | |
Cutout: Left and right side | |
3.2cm | |
100kg | |
182.9cm |
0.4 in. | |
1.0cm | |
Corrosion-resistant epoxy resin | |
1.25 in. | |
220 lb. | |
72 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title