Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Protector™ Downdraft Powder Station

The Labconco Protector™ Downdraft Powder Station keeps users safe by containing powders during fingerprint processing.

Manufacturer:  Labconco™ 3410001

 View more versions of this product

Catalog No. 09-681-305


Add to cart

Alert:

Does not provide protection from biohazardous aerosols or gaseous contaminants. Contact your Fisher Customer Service Representative for information on biosafety cabinets and fume hoods if your application involves these contaminants.


Description

Description

Open-sided, open-top workstation provides unrestricted operator movement and accommodates oversized evidence.

  • Powder-coated steel housing and shelf
  • Type 304 stainless steel work surface with Flow-Zone™ perforations
  • Built-in blower with motorized, backward curved fan and switch
  • With removable powder catch pan
  • Exhaust prefilter of pleated paper with MERV 8 rating
  • Exhaust HEPA filter, 99.99% efficient on particles 0.3 micron
  • Red, filter system alarm light alerts operator to low airflow conditions
  • Comes with 6 ft. (15cm) power cord with plug    
  • Available in 2 ft. (0.6m) and 3 ft. (0.9m) widths and Domestic electrical standards

An optional Transition Connector mounts between two downdraft powder hoods for a secure fit and continuous work surface.

Compliance

ETL listed (Models 3410000 and 3410001).

Specifications

Specifications

115V 60Hz
115V
65.3cm
13A
Stainless Steel
87 lb.
36 in.
25.7 in.
91.4cm
18.75 in.
47.6cm
ETL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit