Learn More
Labconco™ Protector™ Filtered Fume Hood Accessory, Light Bulb
SDP
Replacement light blub for use with Protector Echo and Airo Filtered Fume Hood
Supplier:  Labconco™ 9577700
Catalog No. 10-369-254
Add to Cart
Description
Description
  • Pre-wired vapor-proof fluorescent lighting provides increased visibility in the fume hood itself, up to 63 foot candles
  • Number of bulbs varies based on hood size (check Order Information for quantity needed)
  • First set of bulbs (of number required based on hood width) included with each Protector Echo Filtered Fume Hood
Order Info
Light bulb quantity per model: 3 ft. Hood (1 required); 4 ft. Hood (2 required); 5 ft. Hood (3 required); 6 ft. Hood (4 required); 8 ft. Hood (5 required)
Warranty and Services
One year
Specifications
Specifications
2 lb.
9 in.
Protector Echo and Airo Filtered Fume Hood
5.08 cm
2.54 cm
907.1 g
22.8 cm
2 in.
1 in.
Linear Bulb
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.