Learn More
Labconco™ Protector™ Echo™ Filtered Fume Hood Accessory, Sash Opening Reduction Wing Kits
For use with Protector™ Echo™ Benchtop Filtered Fume Hoods
Manufacturer: Labconco™ 9584904
Description
- Wings reduce sash opening area to increase face velocity at 40.6cm (16 in.) sash stop position from 60fpm to 100fpm
- Hinged with detents to maintain open position for loading and closed position for normal operation
- Wings attach to sides of sash opening
Warranty and Services
One year
Specifications
17 in. | |
4 ft. Protector™ Echo™ Filtered Fume Hood | |
Kit | |
907.1kg | |
20.3cm |
43.1cm | |
Set of 2 wings and mounting hardware | |
2 lb. | |
8 in. |