Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Protector™ Evidence Drying Cabinet: 5 ft.W

Provide a compact, space-saving method of securely drying evidence in your lab with the Labconco Protector Evidence Drying Cabinet. This durable, stainless steel-lined cabinet provides a secure ventilated area to dry evidence such as clothing and bedding and its HEPA and carbon filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors.

Manufacturer:  Labconco™ 3405014

 View more versions of this product

Catalog No. 09-682-757


Add to cart

Description

Description

A built-in blower provides airflow through the cabinet interior to speed drying. Air passes through the door inlet prefilter, circulates up through the interior, passes through the exhaust prefilter, HEPA
and carbon filters and is exhausted back to the room through the top of the cabinet.

  • Durable, stainless steel-lined construction
  • HEPA and carbon-filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors
  • Ultraviolet light provides secondary disinfection of interior between cases to prevent cross contamination, ensuring integrity of samples for DNA testing
  • Lockable cabinet maintains the chain of custody
  • UV security lock limits access to the ultraviolet light switch
  • Safety glass door with magnetic seal maximizes visibility into the cabinet while withstanding exposure to ultraviolet light
  • Inlet roughing prefilter traps dust and other large particles in the air entering the cabinet
  • Storage area below cabinet houses drain bucket and holds miscellaneous cleaning supplies
  • Leak-tight brushed stainless-steel interior cleans easily
  • Three-position switch activates fan or ultraviolet light (only when UV security lock key is in ON position)
  • Spacious interior measures 26.5D x 32.25W x 56 in.H (81.9 x 67.3 x 142.2cm)
  • Stainless-steel hanging rod accommodates clothing, bedding and other tall or bulky items; three wire mesh shelves
  • Exhaust roughing prefilter traps particulates in the exhaust air to prolong the life of the HEPA filter
  • HEPA filter, 99.99% efficient on particles 0.3 micron, traps airborne particulates including biohazardous aerosols, as air is exhausted from cabinet interior
  • Large 14-quart plastic bucket, located in the storage area below the cabinet, collects liquids from the drain in the cabinet's floor to aid clean up

Specifications

Specifications

3A
26.5 in.
19 in.
71.1cm
230V 50/60Hz
HEPA and Carbon filter
79 in.
12 in.
200.7cm
30.48cm
Washdown system with pump
China/Australia
Evidence Drying Cabinet
675 lb.
60 in.
8 in.
152.4cm
20.3cm
Yes
Evidence drying lockable cabinet
28 in.
67.31cm
48.26cm
Safety glass door with magnetic seal
79 in.
12 in.
200.7cm
30.48cm
50/60Hz
Stainless Steel lined
Washdown system with pump
220V
60 in.
306kg
36 in.
152.4cm
91.44cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit